missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Dispensers

Brand:  Duran™ 291151707

Code : 33

Additional Details : Weight : 0.32600kg

 View more versions of this product

Product Code. 13157124

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

5 to 50mL
<±0.6 to 0.5%
DIN ISO 8655
Dispenser
At 121 °C
<± 0.1%
5 to 50mL
GL 32, GL 38, S 40 Adapters
Videos