missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Dispensers

Brand:  Duran™ 291150608

Code : 33

Additional Details : Weight : 0.17400kg

 View more versions of this product

Product Code. 13157604

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

0.5 to 5mL
<±0.6 to 0.5%
DIN ISO 8655
Dispenser
At 121 °C
<± 0.1%
0.5 to 5mL
GL 28, GL 45, S 40 Adapters
Videos