Learn More
Fisherbrand™ Dispenser Tips
Robust, stable, and durable dispenser tips for several brands of pipettors.
Brand:  Fisherbrand™
Code : 41
Additional Details : Weight : 0.21000kg
Product Code. 11562363
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
These tips are for use with the following pipettors:
  • Brand Handystep® and Handystep® electronic All tips
  • Eppendorf® Multipette® (not Multipette® Plus) All tips
  • Gilson Distriman® 
  • Ritter Ripette® All tips
  • Tri-continent Easystep® 
Additonal features:
  • These Dispenser® tips are sterile without an adapter
Specifications
Specifications
Dispenser tip
X100
Beta sterilized
Videos
Documentation Alternative Products