Learn More
3M™ Disc Filters
Brand: 3M™ 7000029735
Code : NEW
Additional Details : Weight : 0.41850kg
Specifications
P3 | |
GP2/GP3 | |
6000, 7000, and 7500 series half and full face masks |
Particulate Filter | |
Protection agains solid and liquid particles up to 20 x WEL (half-mask) or 40 x WEL (full face mask) plus ozone to 10x WEL and organic vapors/acid gas below WEL | |
20/Pk. |