missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Direct Red 80
19.38 GBP valid until 2024-04-26
Use promo code "21687" to get your promotional price.
Use promo code "21687" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Direct red 80 is a dye useful as a cardiac muscle stain and as a model compound in adsorption studies. It has also been utilized to compare efficacy of textile dye removal in aqueous solutions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Direct red 80 is a dye useful as a cardiac muscle stain and as a model compound in adsorption studies. It has also been utilized to compare efficacy of textile dye removal in aqueous solutions.
Solubility
Partly soluble in water, very slightly soluble in ethanol, and cellosolve. Insoluble in many organic solvents.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Direct red 80 is a dye useful as a cardiac muscle stain and as a model compound in adsorption studies. It has also been utilized to compare efficacy of textile dye removal in aqueous solutions.
Solubility
Partly soluble in water, very slightly soluble in ethanol, and cellosolve. Insoluble in many organic solvents.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Molecular Formula | C45H26N10Na6O21S6 |
Solubility Information | Partly soluble in water,very slightly soluble in ethanol,and cellosolve. Insoluble in many organic solvents. |
InChI Key | LDCKXVAPQFUIOI-ZFLDVXHKSA-H |
SMILES | C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C(=CC4=C(C3=O)C=CC(=C4)NC(=O)NC5=CC6=C(C=C5)C(=O)C(=NNC7=C(C=C(C=C7)N=NC8=CC=C(C=C8)S(=O)(=O)[O-])S(=O)(=O)[O-])C(=C6)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+] |
IUPAC Name | hexasodium;4-oxo-7-[[5-oxo-7-sulfonato-6-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalen-2-yl]carbamoylamino]-3-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2-sulfonate |
Molecular Weight (g/mol) | 1373.05 |
PubChem CID | 57369623 |
Formula Weight | 1373.09 |
Quantity | 5 g |
Chemical Name or Material | Direct Red 80 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'