Learn More
DIPPA hydrochloride, Tocris Bioscience™
Selective irreversible κ antagonist
Brand:  Tocris (Bio-Techne) 079450
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13488013
  • £554.00 / 50mg
Estimated Shipment
07-03-2018
Add to basket
Description
Description
An irreversible and selective antagonist at the κ receptor, with persistent effect in vivo.
Specifications
Chemical Identifiers
DIPPA hydrochloride
C22H23Cl2N3OS·HCl
484.87
2-(3, 4-Dichlorophenyl)-N-methyl-N-[(1S)-1-(3-isothiocyanatophenyl)-2-(1-pyrrolidinyl)ethyl]acetamide hydrochloride
Specifications
DIPPA hydrochloride
155512-52-0
C22H23Cl2N3OS·HCl
2-(3, 4-Dichlorophenyl)-N-methyl-N-[(1S)-1-(3-isothiocyanatophenyl)-2-(1-pyrrolidinyl)ethyl]acetamide hydrochloride
50mg
>99%
484.87
Videos