missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenylphosphinic acid, 99%, Thermo Scientific Chemicals
17.66 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Diphenylphosphinic acid is used as a reagent employed in the synthesis of bidentate ligands and peptide coupling agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diphenylphosphinic acid is used as a reagent employed in the synthesis of bidentate ligands and peptide coupling agents.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Diphenylphosphinic acid is used as a reagent employed in the synthesis of bidentate ligands and peptide coupling agents.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 193°C to 197°C |
Quantity | 5 g |
Beilstein | 2804567 |
Solubility Information | Soluble in water. |
Formula Weight | 218.19 |
Percent Purity | 99% |
Chemical Name or Material | Diphenylphosphinic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'