missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Diphenylmercury

Brand:  Alfa Aesar™ 037119.06

Code : NEW

Additional Details : CAS Number : 587-85-9 Weight : 0.00500kg Transport : UN number : 2026 Chem class : 1 Pack group : II

Quantity 5g

 View more versions of this product

Product Code. 11394287

  • £47.20 / 5g


Estimated Shipment date
from Supplier 16-02-2018
Add to basket

Specifications

Chemical Identifiers

Diphenylmercury
354.81
UN2026
587-85-9
MFCD00002998
637234

Specifications

Diphenylmercury
5g
2.318g/mL
Powder
MFCD00002998
637234
124° to 125°C
587-85-9
>306°C (dec.)
354.81
UN2026
Videos
SDS