missing translation for 'onlineSavingsMsg'
Learn More

Diphenyliodonium chloride, 97%, ACROS Organics™

Brand:  Acros Organics 117330500

Code : 29

Additional Details : CAS Number : 1483-72-3 Weight : 0.10000kg Transport : UN number : 2811 Chem class : 1 Pack group : III


Quantity 50g
Packaging Glass bottle

 View more versions of this product

Product Code. 10656734

  • £360.90 / 50g


Estimated Shipment
17-05-2018
Add to basket


Specifications

Chemical Identifiers

Diphenyliodonium chloride
97%
(C6H5)2ICl
MFCD00011909
≥96.0%
C12H10I·Cl
316.57

Specifications

Diphenyliodonium chloride
Authentic
Powder
97%
(C6H5)2ICl
MFCD00011909
White to light cream-yellow
1483-72-3
Glass bottle
≥96.0%
C12H10I·Cl
316.57
01,341; 02,178
50g
Videos
SDS