missing translation for 'onlineSavingsMsg'
Learn More

Diphenyliodonium chloride, 97%, Thermo Scientific Chemicals

Catalog Number p-3778451
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1483-72-3
C12H10ClI
316.57
MFCD00011909
RSJLWBUYLGJOBD-UHFFFAOYSA-M
diphenyliodonium chloride, diphenyliodanium chloride, iodonium, diphenyl-, chloride, diphenyliodoniumchloride, c12h10i.cl, iodonium, diphenyl-, chloride 1:1, acmc-1bxx9
73870
diphenyliodanium;chloride
[Cl-].[I+](C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
1483-72-3
C12H10ClI
316.57
MFCD00011909
RSJLWBUYLGJOBD-UHFFFAOYSA-M
diphenyliodonium chloride, diphenyliodanium chloride, iodonium, diphenyl-, chloride, diphenyliodoniumchloride, c12h10i.cl, iodonium, diphenyl-, chloride 1:1, acmc-1bxx9
73870
diphenyliodanium;chloride
[Cl-].[I+](C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 227°C to 228°C
Color Cream-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (C6H5)2ICl
Quantity 10 g
Fieser 01,341; 02,178
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol
Formula Weight 316.57
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Diphenyliodonium chloride
Show More Show Less
compliance-icons
Product Identifier
  • Diphenyliodonium chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'