missing translation for 'onlineSavingsMsg'
Learn More

Diphenylcarbamyl chloride, 98%, ACROS Organics™

Brand:  Acros Organics 167145000

Code : 29

Additional Details : CAS Number : 83-01-2 Weight : 0.55000kg Transport : UN number : 3261 Chem class : 8 Pack group : II


Quantity 500g
Packaging Glass bottle

 View more versions of this product

Product Code. 10494765

  • £267.90 / 500g


Estimated Shipment
21-06-2018
Add to basket


Specifications

Chemical Identifiers

Diphenylcarbamyl chloride
98%
(C6H5)2NCOCl
MFCD00000633
98%
C13H10ClNO
231.68
Diphenylcarbamoyl chloride

Specifications

Diphenylcarbamyl chloride
Authentic
Crystalline Powder
98%
(C6H5)2NCOCl
231.68
15, 3351
81.0° to 85.0°C
500g
83-01-2
Glass bottle
98%
C13H10ClNO
Solubility in water: reacts.
MFCD00000633
Diphenylcarbamoyl chloride
Beige to gray-green
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Diphenylcarbamyl chloride

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS