missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenylamine, ACS reagent, Thermo Scientific Chemicals
14.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Melting Point | 52.0°C to 54.0°C |
Color | White |
Boiling Point | 302.0°C |
Flash Point | 152°C |
Ignition Residue | 0.03% max. |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Linear Formula | (C6H5)2NH |
Quantity | 5 g |
Beilstein | 12, 174 |
Show More |
Safety and Handling
Product Identifier
- Diphenylamine
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Specific target organ toxicity after repeated exposure Category 2
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- MIXTURE LIST-Contains : Diphenylamine
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'