missing translation for 'onlineSavingsMsg'
Learn More

Diphenylamine, ACS, Thermo Scientific Chemicals

Catalog Number p-7050367
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
122-39-4
C12H11N
169.227
MFCD00003014
DMBHHRLKUKUOEG-UHFFFAOYSA-N
diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald
11487
CHEBI:4640
N-phenylaniline
C1=CC=C(C=C1)NC2=CC=CC=C2
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
122-39-4
C12H11N
169.227
MFCD00003014
DMBHHRLKUKUOEG-UHFFFAOYSA-N
diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald
11487
CHEBI:4640
N-phenylaniline
C1=CC=C(C=C1)NC2=CC=CC=C2

Diphenylamine is employed as harvest scald inhibitor for apples indoor drench treatments due to its antioxidant property. Alkyl derivatives of diphenylamine are used as anti-ozonants in the rubber manufacturing. It is used as lubricants and acts as a stabilizer for plastics. In the analytical chemistry, it is used to test the nitrate ion in association with ammonium chloride in sulfuric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 52°C to 55°C
Density 1.16 g/mL
Boiling Point 302°C
Flash Point 152°C (305°F)
Linear Formula (C6H5)2NH
Quantity 25 g
UN Number UN3077
Beilstein 508755
Sensitivity Air and light sensitive
Merck Index 14,3317
Solubility Information Soluble in ethanol,acetone,benzene,carbon tetrachloride,pyridine,ethyl acetate; Slightly soluble in chloroform.
Formula Weight 169.23
Grade ACS
Physical Form Solid
Chemical Name or Material Diphenylamine, For analysis ACS
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'