missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl diselenide, 99%, Thermo Scientific Chemicals

Catalog Number p-3736384
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1666-13-3
C12H10Se2
312.15
MFCD00003001
YWWZCHLUQSHMCL-UHFFFAOYSA-N
diphenyl diselenide, diselenide, diphenyl, phenyl diselenide, 1,2-diphenyldiselane, diphenyldiselenide, bis phenylselenide, phenyldiselanyl benzene, diphenyldiselane, ccris 9227, acmc-209duq
15460
(phenyldiselanyl)benzene
[Se]([Se]C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
1666-13-3
C12H10Se2
312.15
MFCD00003001
YWWZCHLUQSHMCL-UHFFFAOYSA-N
diphenyl diselenide, diselenide, diphenyl, phenyl diselenide, 1,2-diphenyldiselane, diphenyldiselenide, bis phenylselenide, phenyldiselanyl benzene, diphenyldiselane, ccris 9227, acmc-209duq
15460
(phenyldiselanyl)benzene
[Se]([Se]C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 60.0°C to 63.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)2Se2
Quantity 5 g
Beilstein 06, 346
Fieser 05,272; 06,235; 07,136; 09,199; 10,170; 11,219; 13,125; 16,152; 17,352
Solubility Information Solubility in water: insoluble in water.
Formula Weight 312.13
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Diphenyl diselenide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'