missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenyl cyanocarbonimidate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Diphenyl N-cyanocarbonimidate has been used in the preparation of N-substituted 2-amino-4H-3,1-benzoxazines, cyanoguanidine derivatives, oxadiazoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diphenyl N-cyanocarbonimidate has been used in the preparation of N-substituted 2-amino-4H-3,1-benzoxazines, cyanoguanidine derivatives, oxadiazoles.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep under a nitrogen blanket. Store away from strong oxidizing agents, strong acids, strong bases, moist air, water.
Diphenyl N-cyanocarbonimidate has been used in the preparation of N-substituted 2-amino-4H-3,1-benzoxazines, cyanoguanidine derivatives, oxadiazoles.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep under a nitrogen blanket. Store away from strong oxidizing agents, strong acids, strong bases, moist air, water.
Specifications
Melting Point | 155°C to 158°C |
Linear Formula | (C6H5O)2C=NCN |
Quantity | 1 g |
Sensitivity | Moisture sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 238.25 |
Percent Purity | 97% |
Chemical Name or Material | Diphenyl cyanocarbonimidate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'