missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl-2-pyridylmethane 98%, ACROS Organics™

Brand:  Acros Organics 387940250

Code : 29

Additional Details : CAS Number : 3678-70-4 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10611115

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Diphenyl-2-pyridylmethane
98%
C18H15N
MFCD00006309
3678-70-4
98%
245.32

Specifications

Diphenyl-2-pyridylmethane
54.0° to 61.0°C
3678-70-4
Glass bottle
98%
C18H15N
MFCD00006309
Light beige to yellow-orange
25g
Authentic
Crystalline Powder and Crystals
98%
245.32
2-Diphenylmethylpyridine
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Diphenyl-2-pyridylmethane

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ A§ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS