missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl-2-pyridylmethane 98%, ACROS Organics™

Brand:  Acros Organics 387940050

Code : 29

Additional Details : CAS Number : 3678-70-4 Weight : 0.06000kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10138202

  • £16.30 / 5g


Estimated Shipment
13-02-2018
Add to basket


Specifications

Chemical Identifiers

Diphenyl-2-pyridylmethane
98%
C18H15N
MFCD00006309
3678-70-4
98%
245.32

Specifications

Diphenyl-2-pyridylmethane
54.0° to 61.0°C
3678-70-4
Glass bottle
98%
C18H15N
MFCD00006309
Light beige to yellow-orange
5g
Authentic
Crystalline Powder and Crystals
98%
245.32
2-Diphenylmethylpyridine
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Diphenyl-2-pyridylmethane

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS