missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Dipentene, tech.

Brand:  Alfa Aesar™ 045805.AD

Code : NEW

Additional Details : CAS Number : 138-86-3 Weight : 0.00050kg Transport : UN number : 2052 Chem class : 1 Pack group : III

Quantity 50mL

 View more versions of this product

Product Code. 15412457

  • £13.10 / 50mL


Estimated Shipment date
from Supplier 21-03-2018
Add to basket

Specifications

Chemical Identifiers

Dipentene
Technical Grade
CH3C6H8C(CH3)=CH2
MFCD00062992
3587825
(+/-)-Limonene; (+/-)-p-Mentha-1,8-diene
138-86-3
C10H16
136.24
UN2052
14,5493

Specifications

Dipentene
0.85g/mL
Liquid
CH3C6H8C(CH3)=CH2
136.24
UN2052
14,5493
170° to 180°C
43°C (109°F)
138-86-3
Technical Grade
C10H16
1.474
MFCD00062992
3587825
(+/-)-Limonene; (+/-)-p-Mentha-1,8-diene
50mL
Videos
SDS