missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipentene, pure, fraction of terpene hydrocarbons, Thermo Scientific Chemicals
16.66 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 0.8680g/mL |
Boiling Point | 174.0°C |
Flash Point | 42°C |
Packaging | Glass bottle |
Refractive Index | 1.4730 to 1.4780 (20°C, 589nm) |
Quantity | 1 L |
Specific Gravity | 0.868 |
Formula Weight | 136.23 |
Grade | Pure |
Physical Form | Liquid |
Show More |
Safety and Handling
Product Identifier
- Dipentene
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Aspiration toxicity Category 1
- Flammable liquid Category 3
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- H304-May be fatal if swallowed and enters airways.
- H317-May cause an allergic skin reaction.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P331-Do NOT induce vomiting.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'