Learn More
Thermo Scientific™ Dionex™ Zero Dead Volume Fitting
Thermo Scientific™ Dionex Zero Dead Volume Fittings are for older generation nano and capillary LC systems. For dead volume free connections, consider switching to new nanoViper™ connection technology.
Brand: Thermo Scientific™ 160705
Code : 16
Additional Details : Weight : 0.10000kg
Description
Thermo Scientific™ Dionex Zero Dead Volume Fittings are for older generation nano and capillary LC systems.
Specifications
Zero dead volume fitting | |
nanoLC | |
1/16 in. |
Chromatography | |
Fitting |