missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ OmniPac™ PAX-500 HPLC Columns

When you need to separate anionic and neutral species in a single run, use this solvent-compatible mixed-mode column.

Brand:  Thermo Scientific™ 042152

Code : 16

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 11301723

  • £1227.98 / Each
Estimated Shipment: 02-07-2024
to see stock.
Add to basket

Description

Description

When you need to separate anionic and neutral species in a single run, use the Thermo Scientific™ OmniPac™ PAX-500 Column. With anion-exchange and reversed-phase properties, this polymer-based mixed-mode column is solvent compatible and stable over the entire pH range of 1-14. Solvents may be used to affect selectivity as well as to remove contaminants to extend column lifetime. The OmniPac PAX-500 column is ideal for analysis of a wide range of compounds.

The OmniPac PAX-500 column is ideal for analysis of alcohols, alkanolamines, antihistamines, anti-inflammatory agents, aromatic acids, inorganic acids, peptides, purines and pyrimidines, sulfonamides, sulfonated anionic surfactants, and water-soluble vitamins.
  • Macroporous hydrophobic core provides reversed-phase retention and is coated with an anion-exchange latex bead layer for anion-exchange retention
  • Ion exchange capacity of approximately 40μeq/column
  • 100% solvent-compatible and should always be run with at least 1% organic solvent in the mobile phase
Specifications

Specifications

Mixed Mode
Reversed Phase and Ion Exchange
8.5 μm
Spherical, Microporous Polymer
1200 Å
L50
4 mm
1 mL/min
4000 psi (275 bar)
Alkanol Quaternary Amine
0 to 14
300 m²/g
250 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.