missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Dionex™ Fused Silica Tubing

Thermo Scientific™ Fused Silica Tubing provides excellent chemical durability and inertness.

Brand:  Thermo Scientific™ 163934

Code : 16

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 11312233

  • £72.75 / Each


Estimated Shipment
07-02-2018
Add to basket

Specifications

Specifications

LC-packings Ultimate Capillary/nano HPLC
I.D. 30µm O.D. 375µm
Fused Silica
Capillary Tubing
2.5m
30um I.D.; 375um O.D.; 2.5m length
Videos