missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DNAPac™ PA200 Oligonucleotide Columns

Enjoy strong anion exchange for high-resolution analysis and purification of synthetic oligonucleotides with Thermo Scientific™ DNAPac™ PA200 Oligonucleotide Columns.

Brand:  Thermo Scientific™ SP6734

Code : 16

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 11302503

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

  • Achieve n, n-1 resolution for oligonucleotides
  • Resolve oligonucleotides with secondary structures
  • Assay phosphorothioate purity
  • Selectivity control with eluent pH, salt, and solvent
  • Resolve RNA with aberrant (2' – 5') links from normal SSRNA
Specifications

Specifications

UltiMate 3000 Titanium System
Analytical Column
N/A
1.2mL/min.
pH 2-12.5, 100% compatible with common organic solvents, Ionic form eluents: chloride, perchlorate
8µm
Non-porous
-
Nucleic Acid/Anion Exchange
Analytical Column
DNAPac PA200
Ambient - 85°C
Ion Exchange
-
-
4000psi
Spherical, non-porous polymer
85°C
2.5 to 12.5
250mm
PEEK
8µm diameter nonporous substrate (55% cross-linking) agglomerated with a 130nm MicroBead™ alkyl quaternary ammonium functionalized latex (5% cross-linked)
22mm
Videos

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.