missing translation for 'onlineSavingsMsg'
Learn More

Dimethylzinc, 96%, Thermo Scientific Chemicals

Catalog Number 15491537
Click to view available options
Quantity:
25 g
Packaging:
Cylinder
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 544-97-8
Melting Point -40°C
Density 1.386 g/mL at 20°C
Boiling Point 46°C
Flash Point -18°C (-0.4°F)
Packaging Cylinder
Molecular Formula C2H6Zn
MDL Number MFCD00014854
Quantity 25 g
UN Number UN3394
Beilstein 3587195
Sensitivity Air and moisture sensitive
Merck Index 14,3263
Synonym dimethylzinc, zinc, dimethyl, dimethyl zinc, methylzinc, unii-8h6r0n8w2f, ch3 2zn, znme2, zinc methyl, dimethylzink, hsdb 709
InChI Key JRPGMCRJPQJYPE-UHFFFAOYSA-N
SMILES [CH3-].[CH3-].[Zn+2]
IUPAC Name zinc;carbanide
Molecular Weight (g/mol) 95.45
PubChem CID 11010
Formula Weight 95.44
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Dimethylzinc
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'