Learn More
Dimethylzinc, 1.2M solution in toluene, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 377248000
574.50 GBP valid until 2024-07-31
Use promo code "23251" to get your promotional price.
Code : 29
Additional Details : CAS Number : 544-97-8 Weight : 0.74400kg Transport : UN number : 3399 Chem class : 4.3 Pack group : I
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dimethylzinc | |
1.2M Solution in Toluene | |
C2H6Zn | |
MFCD00014854 | |
0.9300g/mL | |
dimethylzinc, zinc, dimethyl, dimethyl zinc, methylzinc, unii-8h6r0n8w2f, ch3 2zn, znme2, zinc methyl, dimethylzink, hsdb 709 | |
JRPGMCRJPQJYPE-UHFFFAOYSA-N | |
zinc;carbanide | |
11010 | |
AcroSeal™ Glass Bottle | |
800 mL |
Clear colorless | |
544-97-8,108-88-3 | |
Zn(CH3)2 | |
1.2M (based on Zn) exact strength on the certificate of analysis | |
15, 3289 | |
Solubility in water: vigorous reaction. Other solubilities: soluble in hydrocarbons | |
[CH3-].[CH3-].[Zn+2] | |
95.45 | |
95.45 | |
0.93 |
RUO – Research Use Only