missing translation for 'onlineSavingsMsg'
Learn More

Dimethyltin oxide, Thermo Scientific Chemicals

Catalog Number p-7080329
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
2273-45-2
C2H6OSn
164.78
MFCD00013838
WNVQCJNZEDLILP-UHFFFAOYSA-N
dimethyltin oxide, stannane, dimethyloxo, dimethyloxostannane, tin, dimethyloxo, dimethyltinoxide, dimethyl oxo tin, dimethyl tin oxide, unii-409qf23wo7, dimethyl oxo stannane, oxodimethyltin
75277
dimethyl(oxo)tin
C[Sn](C)=O
This item is not returnable. View return policy
2273-45-2
C2H6OSn
164.78
MFCD00013838
WNVQCJNZEDLILP-UHFFFAOYSA-N
dimethyltin oxide, stannane, dimethyloxo, dimethyloxostannane, tin, dimethyloxo, dimethyltinoxide, dimethyl oxo tin, dimethyl tin oxide, unii-409qf23wo7, dimethyl oxo stannane, oxodimethyltin
75277
dimethyl(oxo)tin
C[Sn](C)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point (decomposition)
Quantity 2 g
UN Number UN3146
Formula Weight 166.77
Physical Form Powder
Chemical Name or Material Dimethyltin oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'