missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl sulfate, 99+%, AcroSeal™, Thermo Scientific Chemicals
13.23 GBP valid until 2024-03-30
Use promo code "21636" to get your promotional price.
16.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
13.80 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21636" to get your promotional price.
16.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
13.80 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
Specifications
Melting Point | -32.0°C |
Density | 1.3300g/mL |
Boiling Point | 188.0°C |
Flash Point | 83°C |
Assay Percent Range | 99+% |
Packaging | AcroSeal™ Glass Bottle |
Linear Formula | (CH3)2SO4 |
Refractive Index | 1.3860 to 1.3880 |
Quantity | 100 mL |
Merck Index | 15,328 |
Show More |
Safety and Handling
Product Identifier
- Dimethyl sulfate
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- H317-May cause an allergic skin reaction.
- H314-Causes severe skin burns and eye damage.
- H350-May cause cancer.
- H330-Fatal if inhaled.
- H341-Suspected of causing genetic defects.
- H301-Toxic if swallowed.
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'