missing translation for 'onlineSavingsMsg'
Learn More

Dimethylphosphoramidous Dichloride, 97%, ACROS Organics™

Brand:  Acros Organics 383350250

Code : 29

Additional Details : CAS Number : 683-85-2 Weight : 0.08178kg Transport : UN number : 2924 Chem class : 3 Pack group : II


Quantity 25mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10300983

  • £145.90 / 25mL


EU Stock 3
Add to basket


Specifications

Chemical Identifiers

Dimethylphosphoramidous dichloride
C2H6Cl2NP
145.96
97%
(CH3)2NPCl2
MFCD00013613

Specifications

Dimethylphosphoramidous dichloride
−23°C
Colorless to light yellow
683-85-2
96.0% min.
1.5040 to 1.5060 (20°C, 589nm)
97%
C2H6Cl2NP
145.96
1.2710g/mL
150.0°C
25mL
Authentic
Glass bottle
Liquid
97%
(CH3)2NPCl2
MFCD00013613
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Dimethylphosphoramidous dichloride

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS