missing translation for 'onlineSavingsMsg'
Learn More

Dimethylphosphoramidous Dichloride, 97%, Thermo Scientific Chemicals

Catalog Number p-3212644
Click to view available options
Quantity:
5 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
683-85-2
C2H6Cl2NP
145.96
MFCD00013613
XPWWDZRSNFSLRQ-UHFFFAOYSA-N
dimethylphosphoramidous dichloride, dichloro dimethylamino phosphine, phosphoramidous dichloride, dimethyl, dimethylamino dichlorophosphine, dimethylphosphoramidousdichloride, me2npcl2, dimethylaminodichlorphosphine, dimethylaminodichlorophosphine, ch3 2npcl2, dichlorophosphanyl dimethylamine
136483
N-dichlorophosphanyl-N-methylmethanamine
CN(C)P(Cl)Cl
This item is not returnable. View return policy
683-85-2
C2H6Cl2NP
145.96
MFCD00013613
XPWWDZRSNFSLRQ-UHFFFAOYSA-N
dimethylphosphoramidous dichloride, dichloro dimethylamino phosphine, phosphoramidous dichloride, dimethyl, dimethylamino dichlorophosphine, dimethylphosphoramidousdichloride, me2npcl2, dimethylaminodichlorphosphine, dimethylaminodichlorophosphine, ch3 2npcl2, dichlorophosphanyl dimethylamine
136483
N-dichlorophosphanyl-N-methylmethanamine
CN(C)P(Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.2710g/mL
Boiling Point 150.0°C
Flash Point −23°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CH3)2NPCl2
Refractive Index 1.5040 to 1.5060 (20°C, 589nm)
Quantity 5 mL
Specific Gravity 1.271
Solubility Information Solubility in water: reacts
Formula Weight 145.96
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dimethylphosphoramidous dichloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dimethylphosphoramidous dichloride
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'