missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylglyoxime, Pure, Fisher Chemical™
Click to view available options
Quantity:
25 g
100 g
Specifications
Melting Point | 239°C |
Color | White |
Packaging | Plastic powder jar |
Quantity | 25 g |
Formula Weight | 116.12g/mol |
Physical Form | Solid |
Safety and Handling
Product Identifier
- Dimethylglyoxime
- Danger
- Flammable solid Category 2
- Acute toxicity Category 3
- H228-Flammable solid.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'