missing translation for 'onlineSavingsMsg'
Learn More

Dimethylglyoxime, ACS, 99+%, Thermo Scientific Chemicals

Catalog Number p-7050015
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
95-45-4
C4H8N2O2
116.12
MFCD00002117
JGUQDUKBUKFFRO-GGWOSOGESA-N
biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky
5356010
N-[(E)-3-nitrosobut-2-en-2-yl]hydroxylamine
C\C(=N/O)\C(\C)=N\O
This item is not returnable. View return policy
95-45-4
C4H8N2O2
116.12
MFCD00002117
JGUQDUKBUKFFRO-GGWOSOGESA-N
biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky
5356010
N-[(E)-3-nitrosobut-2-en-2-yl]hydroxylamine
C\C(=N/O)\C(\C)=N\O

Dimethylglyoxime, is used as a chelating agent which forms complexes with metals such as copper, nickel, and palladium. Dimethylglyoxime reacts with a nickel salt in a green solution to generate a red colored insoluble tetradentate coordinate complex of nickel. It is used as a reagent for the colorimetric determination of urea and ureido compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼240°C (decomposition)
Color White
Quantity 50 g
Beilstein 506731
Merck Index 14,3246
Solubility Information Soluble in alcohol, acetone, ether
Formula Weight 116.12
Grade ACS
Physical Form Powder
Chemical Name or Material Dimethylglyoxime

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'