missing translation for 'onlineSavingsMsg'
Learn More

Dimethylgermanium dichloride, 97+%, Thermo Scientific Chemicals

Catalog Number p-7050055
Click to view available options
Quantity:
0.5 g
2 g
10 g
This item is not returnable. View return policy
1529-48-2
C2H6Cl2Ge
173.6
MFCD00013588
YQECBLVSMFAWIZ-UHFFFAOYSA-N
dimethylgermanium dichloride, germane, dichlorodimethyl, dimethyldichlorogermane, dichloro dimethyl germane, dimethylgermanedichloride, dimethylgermyl dichloride, geme2cl2, acmc-1boyh, germanium dimethyl dichloride, dimethyl germanium dichloride
73724
dichloro(dimethyl)germane
C[Ge](C)(Cl)Cl
This item is not returnable. View return policy
1529-48-2
C2H6Cl2Ge
173.6
MFCD00013588
YQECBLVSMFAWIZ-UHFFFAOYSA-N
dimethylgermanium dichloride, germane, dichlorodimethyl, dimethyldichlorogermane, dichloro dimethyl germane, dimethylgermanedichloride, dimethylgermyl dichloride, geme2cl2, acmc-1boyh, germanium dimethyl dichloride, dimethyl germanium dichloride
73724
dichloro(dimethyl)germane
C[Ge](C)(Cl)Cl

Dimethylgermanium dichloride is a useful organogermanium compound for synthesis and proteomics research. It is also used as an organic intermediate and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -22°C
Density 1.51 g/mL
Boiling Point 124°C
Flash Point 32°C (90°F)
Refractive Index 1.46
Quantity 0.5 g
UN Number UN2924
Sensitivity Moisture sensitive
Solubility Information Hydrolyses in water.
Formula Weight 173.61
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Dimethylgermanium dichloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'