missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DimethylFormamide, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Plastic Container
Description
DimethylFormamide, Extra Pure, SLR
Specifications
pH | 6 to 8 |
Melting Point | -61°C |
Color | Colorless |
Molecular Weight (g/mol) | 73.10 |
Formula Weight | 73.09g/mol |
Boiling Point | 153°C |
Quantity | 25 L |
Packaging | Plastic Container |
Physical Form | Liquid |
Vapor Pressure | 4.9mbar at 20°C |
Show More |
Safety and Handling
Product Identifier
- Dimethylformamide
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- H226-Flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H360D-May damage the unborn child.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'