missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylethoxysilane, 94%, Thermo Scientific Chemicals
121.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Color | Colorless |
Boiling Point | 54.0°C |
Flash Point | −5°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 92% min. (GC) |
Packaging | Glass bottle |
Linear Formula | CH3CH2OSiH(CH3)2 |
Refractive Index | 1.3670 to 1.3690 (20°C, 589nm) |
Quantity | 100 g |
Solubility Information | Solubility in water: reacts |
Show More |
Safety and Handling
Product Identifier
- Dimethylethoxysilane
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'