missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 4,5-imidazoledicarboxylate, 98%, Acros Organics™

Brand:  Acros Organics 408370250

Code : 29

Additional Details : CAS Number : 3304-70-9 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10500774

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

Dimethyl 4,5-imidazoledicarboxylate
98%
C7H8N2O4
MFCD00051338
3304-70-9
98%
184.15

Specifications

Dimethyl 4,5-imidazoledicarboxylate
197.0° to 201.0°C
3304-70-9
Glass bottle
98%
C7H8N2O4
MFCD00051338
White to light brown
25g
Authentic
Fine Crystalline Powder
98%
184.15
Videos
SDS