missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl (S)-(-)-malate, 98%, Thermo Scientific Chemicals

Catalog Number p-4323524
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
617-55-0
C6H10O5
162.14
MFCD00066215
YSEKNCXYRGKTBJ-UHFFFAOYNA-N
s-dimethyl 2-hydroxysuccinate, dimethyl s---malate, dimethyl l-malate, l---malic acid dimethyl ester, 1,4-dimethyl 2s-2-hydroxybutanedioate, butanedioic acid, hydroxy-, dimethyl ester, 2s, dimethyl l---malate, pubchem6778, di-methyl s-malate, dimethyl s--malate
10285815
dimethyl (2S)-2-hydroxybutanedioate
COC(=O)CC(O)C(=O)OC
This item is not returnable. View return policy
617-55-0
C6H10O5
162.14
MFCD00066215
YSEKNCXYRGKTBJ-UHFFFAOYNA-N
s-dimethyl 2-hydroxysuccinate, dimethyl s---malate, dimethyl l-malate, l---malic acid dimethyl ester, 1,4-dimethyl 2s-2-hydroxybutanedioate, butanedioic acid, hydroxy-, dimethyl ester, 2s, dimethyl l---malate, pubchem6778, di-methyl s-malate, dimethyl s--malate
10285815
dimethyl (2S)-2-hydroxybutanedioate
COC(=O)CC(O)C(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.2300g/mL
Boiling Point 91.0°C to 93.0°C (1.0 Torr)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H3COOCCH(OH)CH2COOCH3
Refractive Index 1.4366
Quantity 5 g
Specific Gravity 1.23
Specific Rotation − 6.70
Solubility Information Solubility in water: in water:(20 °C) insoluble.
Formula Weight 162.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Dimethyl (S)-(-)-malate
Show More Show Less
compliance-icons
Product Identifier
  • Dimethyl (S)-(-)-malate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'