missing translation for 'onlineSavingsMsg'
Learn More

Dimethyldichlorosilane, Pure, Fisher Chemical™

Catalog Number 10498590
Click to view available options
Quantity:
100 mL
250 mL
This item is not returnable. View return policy
75-78-5
C2H6Cl2Si
129.055
491
LIKFHECYJZWXFJ-UHFFFAOYSA-N
dimethyldichlorosilane, silane, dichlorodimethyl, dichloro dimethyl silane, inerton aw-dmcs, dichlorodimethylsilicon, dimethyl-dichlorsilan, inerton dw-dmc, repel-silan, dimethylsilane dichloride, dimethyl dichlorosilane
6398
dichloro(dimethyl)silane
C[Si](C)(Cl)Cl
This item is not returnable. View return policy
75-78-5
C2H6Cl2Si
129.055
491
LIKFHECYJZWXFJ-UHFFFAOYSA-N
dimethyldichlorosilane, silane, dichlorodimethyl, dichloro dimethyl silane, inerton aw-dmcs, dichlorodimethylsilicon, dimethyl-dichlorsilan, inerton dw-dmc, repel-silan, dimethylsilane dichloride, dimethyl dichlorosilane
6398
dichloro(dimethyl)silane
C[Si](C)(Cl)Cl
Melting Point -76°C
Boiling Point 70°C
Packaging Plastic Coated Glass bottle
Quantity 100 mL
Formula Weight 129.06g/mol
Vapor Pressure 150 hPa at 20°C
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dichlorodimethylsilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'