missing translation for 'onlineSavingsMsg'
Learn More

Dimethylcyanamide, 97%, Thermo Scientific Chemicals

Catalog Number p-116861
Click to view available options
Quantity:
5 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
1467-79-4
C3H6N2
70.10
MFCD00001767
OAGOUCJGXNLJNL-UHFFFAOYSA-N
dimethyl cyanamide, cyanamide, dimethyl, n-cyano-n-methylmethanamine, dimethylkyanamid, cyanodimethylamine, n-cyanodimethylamine, dimethylkyanamid czech, unii-6p25ipq8gn, ccris 5909, cyanamide, n,n-dimethyl
15112
dimethylcyanamide
CN(C)C#N
This item is not returnable. View return policy
1467-79-4
C3H6N2
70.10
MFCD00001767
OAGOUCJGXNLJNL-UHFFFAOYSA-N
dimethyl cyanamide, cyanamide, dimethyl, n-cyano-n-methylmethanamine, dimethylkyanamid, cyanodimethylamine, n-cyanodimethylamine, dimethylkyanamid czech, unii-6p25ipq8gn, ccris 5909, cyanamide, n,n-dimethyl
15112
dimethylcyanamide
CN(C)C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -41.0°C
Color Colorless to Yellow
Density 0.8670g/mL
Boiling Point 161.0°C to 163.0°C
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CH3)2NCN
Refractive Index 1.4090 to 1.4110
Quantity 5 mL
Beilstein 04, 74
Specific Gravity 0.867
Solubility Information Solubility in water: miscible. Other solubilities: soluble in acetone,ethyl ether,ethnaol
Formula Weight 70.09
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dimethylcyanamide, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dimethylcyanamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Flammable liquid Category 3
Hazard Statement
  • H310-Fatal in contact with skin.
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'