missing translation for 'onlineSavingsMsg'
Learn More

Dimethylanilinium Tetrakis (pentafluorophenyl)borate, 98%, Thermo Scientific Chemicals

Catalog Number 10611325
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
118612-00-3
C32H12BF20N
801.23
MFCD01074420
BRHZQNMGSKUUMN-UHFFFAOYSA-O
dimethylanilinium tetrakis pentafluorophenyl borate, unii-h8r86l92nd, n,n-dimethylanilinium tetrakis pentafluorophenyl borate, n,n-dimethylbenzenaminium tetrakis perfluorophenyl borate, n,n-dimethylanilinium tetra pentafluorophenyl borate, benzene, pentafluoro-, boron complex, n,n-dimethylanilinium tetrakis pentafluorophenyl borate 1-, benzenamine, n,n-dimethyl-, tetrakis pentafluorophenyl borate 1-, dimethyl phenyl ammonium tetrakis 2,3,4,5,6-pentafluorophenyl borate, borate 1-, tetrakis pentafluorophenyl-, hydrogen, compd. with n,n-dimethylbenzenamine 1:1:1
10996402
dimethyl(phenyl)azanium;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
[B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.C[NH+](C)C1=CC=CC=C1
118612-00-3
C32H12BF20N
801.23
MFCD01074420
BRHZQNMGSKUUMN-UHFFFAOYSA-O
dimethylanilinium tetrakis pentafluorophenyl borate, unii-h8r86l92nd, n,n-dimethylanilinium tetrakis pentafluorophenyl borate, n,n-dimethylbenzenaminium tetrakis perfluorophenyl borate, n,n-dimethylanilinium tetra pentafluorophenyl borate, benzene, pentafluoro-, boron complex, n,n-dimethylanilinium tetrakis pentafluorophenyl borate 1-, benzenamine, n,n-dimethyl-, tetrakis pentafluorophenyl borate 1-, dimethyl phenyl ammonium tetrakis 2,3,4,5,6-pentafluorophenyl borate, borate 1-, tetrakis pentafluorophenyl-, hydrogen, compd. with n,n-dimethylbenzenamine 1:1:1
10996402
dimethyl(phenyl)azanium;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
[B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.C[NH+](C)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Blue-White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Formula Weight 801.23
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Dimethylanilinium tetrakis (pentafluorophenyl)borate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dimethylanilinium tetrakis (pentafluorophenyl)borate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H351-Suspected of causing cancer.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'