missing translation for 'onlineSavingsMsg'
Learn More

Dimethylammonium-dimethylcarbamate, ratio carbon dioxide/dimethylamine <2:1, Thermo Scientific Chemicals

Catalog Number p-115922
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4137-10-4
C5H14N2O2
134.18
MFCD00029888
JIYXHCMRGZVYMA-UHFFFAOYSA-N
DIMCARB
dimethylamine; dimethylcarbamic acid
CNC.CN(C)C(O)=O
This item is not returnable. View return policy
4137-10-4
C5H14N2O2
134.18
MFCD00029888
JIYXHCMRGZVYMA-UHFFFAOYSA-N
DIMCARB
dimethylamine; dimethylcarbamic acid
CNC.CN(C)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 60°C to 61°C
Flash Point 13°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula ((CH3)2NH2)·(CH3)2NCO2)
Refractive Index 1.4520 to 1.456
Quantity 25 g
Specific Gravity 1.05
Solubility Information Solubility in water: may decompose. Other solubilities: forming carbon dioxide,dimethylamine
Viscosity 84.3 mPa.s (20°C)
Formula Weight 134.18
Physical Form Liquid
Chemical Name or Material Dimethylammonium-dimethylcarbamate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dimethylammonium-dimethylcarbamate, ratio carbon dioxide/dimethylamine <2.0/1
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'