missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylammonium-dimethylcarbamate, ratio carbon dioxide/dimethylamine <2:1, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
Specifications
Color | Colorless to Yellow |
Density | 1.0500g/mL |
Boiling Point | 60°C to 61°C |
Flash Point | 13°C |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Linear Formula | ((CH3)2NH2)·(CH3)2NCO2) |
Refractive Index | 1.4520 to 1.456 |
Quantity | 500 g |
Specific Gravity | 1.05 |
Show More |
Safety and Handling
Product Identifier
- Dimethylammonium-dimethylcarbamate, ratio carbon dioxide/dimethylamine <2.0/1
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'