missing translation for 'onlineSavingsMsg'
Learn More

Dimethylammonium-dimethylcarbamate, ratio carbon dioxide/dimethylamine <2:1, ACROS Organics™

Brand:  Acros Organics 292901000

Code : 29

Additional Details : CAS Number : 4137-10-4 Weight : 0.15000kg Transport : UN number : 2733 Chem class : 1b Pack group : II


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10730571

  • £56.40 / 100g


Estimated Shipment
18-05-2018
Add to basket


Description

Description

Phase transfer catalyst
Specifications

Chemical Identifiers

Dimethylammonium-dimethylcarbamate
((CH3)2NH2)·(CH3)2NCO2)
MFCD00029888
C5H14N2O2
134.18
DIMCARB

Specifications

carbon dioxide/dimethylamine: < 2.0/1
1.0500g/mL
100g
Glass bottle
Liquid
0.5% max.
134.18
DIMCARB
Dimethylammonium-dimethylcarbamate
Clear colorless to light yellow
Authentic
1.4520 to 1.4560 (20°C, 589nm)
C5H14N2O2
((CH3)2NH2)·(CH3)2NCO2)
MFCD00029888
Videos
SDS