Learn More
Dimethylamine, 26 wt.% solution in water, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros
Code : NEW
Additional Details : CAS Number : 124-40-3 Weight : 5.00000kg Transport : UN number : 1160 Chem class : 3 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dimethylamine | |
-37.0°C | |
0.8900g/mL | |
−18°C | |
1.37 | |
MFCD00008288 | |
04,39 | |
0.89 | |
dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution | |
ROSDSFDQCJNGOL-UHFFFAOYSA-N | |
N-methylmethanamine | |
674 | |
45.07 | |
Liquid |
124-40-3,7732-18-5 | |
Colorless | |
54.0°C | |
C2H7N | |
(CH3)2NH | |
5 kg | |
07,119 | |
15,325 | |
Solubility in water: >500g/L (20°C). Other solubilities: soluble in alcohol and ether | |
CNC | |
45.07 | |
CHEBI:17170 | |
25 to 27% |
RUO – Research Use Only