missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide, Multi-Compendial, J.T.Baker™

Brand:  JT Baker 9033-07

UNSPSC : 12000000

Additional Details : Weight : 22.50000kg

Quantity 19L
Packaging Poly Pail

 View more versions of this product

Product Code. 15548504

  • £7799.80 / 19 L


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Chemical Identifiers

Dimethyl Sulfoxide
Multi-Compendial
78.13
67-68-5
(CH3)2SO

Specifications

Dimethyl Sulfoxide
Multi-Compendial
78.13
1L = 1.10kg
19L
67-68-5
(CH3)2SO
Poly Pail
89.00°C