missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide, Extra Pure, SLR, Fisher Chemical™

Catalog Number 10080110
64.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


35.76 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.


64.38 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.


Click to view available options
Quantity:
500 mL
2.5 L
5 L
25 L
Packaging:
HDPE Bottle
Plastic Container
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
679
CHEBI:28262
methylsulfinylmethane
CS(C)=O
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
679
CHEBI:28262
methylsulfinylmethane
CS(C)=O

C2H6OS, CAS Number-67-68-5, demsodrox, methyl sulfoxide, dimexide, methane, sulfinylbis, dimethyl sulfoxide, dmso, demavet, dimethylsulfoxide, dimethyl sulphoxide, demasorb, 500mL, 189 deg.C, CHEBI:28262, Colorless, 78.13g/mol, IAZDPXIOMUYVGZ-UHFFFAOYSA-N, methylsulfinylmethane, 18.4 deg.C, 2089

TRUSTED_SUSTAINABILITY
Melting Point 18.4°C
Color Colorless
Boiling Point 189°C
Quantity 500 mL
Packaging HDPE Bottle
Physical Form Liquid
Vapor Pressure 0.55mbar at 20°C
Viscosity 1.98 mPaS at 25°C
Formula Weight 78.13g/mol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'