missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide-d{6}, 99.9% (Isotopic), Thermo Scientific Chemicals

Catalog Number p-7050334
Click to view available options
Quantity:
5 x 1 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
2206-27-1
C2H6OS
84.17
MFCD00002090
IAZDPXIOMUYVGZ-WFGJKAKNSA-N
dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide
75151
[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
2206-27-1
C2H6OS
84.17
MFCD00002090
IAZDPXIOMUYVGZ-WFGJKAKNSA-N
dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide
75151
[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]

Deuterated DMSO is a common solvent used in NMR spectroscopy. It is in combination with pyridine-d5 plays an important role for solution-state 2D NMR of ball-milled plant cell wall gel. Low-energy collisions of ionized acetone-d6, DMSO-d6 and pyridine-d5 with alkanethiolate is used to prepare self-assembled monolayer surfaces of gold.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Density 1.18 g/mL
Flash Point 88°(190°F)
Refractive Index 1.476
Sensitivity Hygroscopic
Quantity 5 x 1 g
Melting Point 20°C
Boiling Point 190°C
Assay Percent Range 99.9% (Isotopic)
Solubility Information Miscible with water and diethyl ether.
IUPAC Name trideuterio(trideuteriomethylsulfinyl)methane
Formula Weight 84.17
Percent Purity 99.9%
Chemical Name or Material Dimethyl Sulfoxide, Isotopic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'