missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number 15578544
Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
This item is not returnable. View return policy
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
679
CHEBI:28262
CS(C)=O
This item is not returnable. View return policy
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
679
CHEBI:28262
CS(C)=O

High quality chemicals for laboratory and specialized industrial use

IUPAC Name methylsulfinylmethane
Formula Weight 78.13
Grade Meets ACS reagent chemical requirements
Quantity 500 mL
Flash Point 89.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.10kg
Chemical Name or Material Dimethyl Sulfoxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'