missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl suberate, 99%, ACROS Organics™

Catalog Number p-120629
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1732-09-8
C10H18O4
202.25
MFCD00008471
LNLCRJXCNQABMV-UHFFFAOYSA-N
dimethyl suberate, octanedioic acid, dimethyl ester, suberic acid, dimethyl ester, octanedioic acid dimethyl ester, suberic acid dimethyl ester, octanedioic acid dimethyl, 1,8-dimethyl octanedioate, octanedioic acid, 1,8-dimethyl ester, dimethyl octane-1,8-dioate
15611
CHEBI:81345
1,8-dimethyl octanedioate
COC(=O)CCCCCCC(=O)OC
This item is not returnable. View return policy
1732-09-8
C10H18O4
202.25
MFCD00008471
LNLCRJXCNQABMV-UHFFFAOYSA-N
dimethyl suberate, octanedioic acid, dimethyl ester, suberic acid, dimethyl ester, octanedioic acid dimethyl ester, suberic acid dimethyl ester, octanedioic acid dimethyl, 1,8-dimethyl octanedioate, octanedioic acid, 1,8-dimethyl ester, dimethyl octane-1,8-dioate
15611
CHEBI:81345
1,8-dimethyl octanedioate
COC(=O)CCCCCCC(=O)OC
Color Colorless to Yellow
Density 1.01g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3O2C(CH2)6CO2CH3
Refractive Index 1.4325 to 1.4345 (20°C, 589nm)
Quantity 25 g
Beilstein 02, 693
Formula Weight 202.25
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Dimethyl suberate, 99%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'