missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl suberate, 99%, ACROS Organics™
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Specifications
Color | Colorless to Yellow |
Density | 1.0100g/mL |
Boiling Point | 268.0°C |
Flash Point | >110°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Linear Formula | CH3O2C(CH2)6CO2CH3 |
Refractive Index | 1.4325 to 1.4345 |
Quantity | 100 g |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'