Learn More
Alfa Aesar™ Dimethyl pyrimidine-2,5-dicarboxylate, 97%
Brand: Alfa Aesar™ H64903.MD
Code : NEW
Additional Details : CAS Number : 115600-17-4 Weight : 0.00025kg
Quantity | 250mg |
---|
Description
NotesStore in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
Dimethyl pyrimidine-2,5-dicarboxylate | |
196.16 | |
23483097 |
115600-17-4 | |
MFCD26127433 | |
Pyrimidine-2,5-dicarboxylic acid dimethyl ester |
Specifications
Dimethyl pyrimidine-2,5-dicarboxylate | |
250mg | |
Powder | |
196.16 | |
23483097 |
White | |
115600-17-4 | |
97% | |
MFCD26127433 | |
Pyrimidine-2,5-dicarboxylic acid dimethyl ester |